Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pipet Brush
SCI_ED

Catalog No. S30424
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S30424 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30424 Supplier C&A Scientific Co Inc Supplier No. BR630
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Brush portion is centered on wire for back-and-forth action.

  • Soft white nylon bristles on No. 23 galvanized wire
  • Dimensions: 0.5 × 3 in. (1.3 × 7.6cm)
  • Overall length: 24 in. (61cm)

Specifications

Product Type Soft Bristled Pipet Brush
Handle Material Galvanized Wire
Color White
Length (English) 24 in.
Length (Metric) 61 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.