Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Pipet Bulb

Catalog No. 16800121 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-800-121 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-800-121 Supplier Bel-Art™ Supplier No. 660000021
Get NMR Transfer Pipettes & Bulbs with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Squeeze the bulb to dispense or retrieve sample liquid. Bel-Art SP Scienceware™ Wilmad™ Pipet Bulb is constructed of durable latex.

  • Latex rubber
  • 6mm dia. bore

Specifications

Length (Metric) 34.93 cm
Material Latex Rubber
Description Latex bulb
Diameter (Metric) Inner 6 mm
Quantity 50 Pack
Model 804
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.