Learn More
Sartorius Pipet Charging Stand and Carousel
GSA_VA
Catalog No. 14559393 Shop All Sartorius Products
Change view
Click to view available options
Number of Channels:
1
4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Number of Channels
14-559-393 4
14-559-392 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-559-393 Supplier Sartorius Supplier No. 730991
Add to Cart
Edge
Add to Cart
Includes
Universal AC-adaptor
Saves bench space. Sartorius Pipet Charging Stand and Carousel holds and stores pipets in an upright position to avoid contamination. For use with PICUS™ and eLINE™ mechanical and electronic pipets.
For storing and charging Picus and eLINE mechanical and electronic pipets when not in use. Pipets should be stored in an upright position in order to avoid contamination from desks and benches.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipette Tips
Material Polypropylene
Number of Channels 4
Type Pipette Holder
For Use With (Equipment) Picus and eLINE Electronic Pipettes
Includes Universal AC adapter