Learn More
Hach Company Pipet, Serological, Sterile, Polystyrene with Plug, 10 mL, 50/pk
Supplier: Hach Company 2092828
Sterile, disposable plastic pipet with plug. Bulk packaged in 25/pack Color coded. Black ascending/descending graduations. Calibrated “To Deliver”. Accuracy ±2%.