Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Pipette Stands
- Stores pipettes in an upright position while not in use to avoid contamination from the desktop
- Linear Stand is designed for all mechanical devices and stores them neatly in a line for instant access
- Carousel stand is ideal for saving bench space and stores up to 6 mechanical pipettes
- mLINE/Proline Plus Single Holder compactly holds a single mechanical pipette
- Colored caps help identify the volume of mLINE pipettes so it can easily be matched to the corresponding tip size
- Includes 5 color-coded replacement caps
- Calibration Tool for mLINE or Proline Plus mechanical pipettes for continuous accuracy and precision
- Can also be used as a tube opener
Specifications
Specifications
| Color | White/Black |
| For Use With | Pipettes |
| Material | Polypropylene |
| Type | Pipette Holder |
| For Use With (Application) | Storing pipets when not in use |
| For Use With (Equipment) | mLINE/Proline Plus Mechanical Pipette |
| Quantity | 1 Each |
| Dimensions (L x W x H) | 29mmW x 250mmD x 60mmH |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?