Learn More
Hach Company Pipet, Volumetric, Class A, Certified, 25 mL, 6/pk
Supplier:  Hach Company 2636170
Encompass_Preferred
Glass Class A pipets serialized and calibrated “To Deliver”. Each comes with Certificate of Identification and Capacity. Marked with an “A” to designate compliance to ASTM E969.
Catalog No. 50-199-6410
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.