Description
- Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
- Tip holder and tip ejector are autoclavable to avoid cross-contamination
- Each pipet bears engraved identification number for traceability
Specifications
Specifications
| Accuracy | ±0.5, ±1 μL |
| Compatible Tips | D200, D300, DF100ST, DF200ST, DF300ST |
| Material | PVDF, Stainless Steel |
| Model | P200 |
| Product Line | Pipetman Classic |
| Type | Pipette |
| Volume (Metric) | 50 to 200 μL |
| For Use With (Application) | Small volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology) |