Learn More
Gilson™ PIPETMAN Classic™ Pipets PROMO
Catalog No. F123615GPR Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
2 to 20 μL
20 to 100 μL
200 to 1000 μL
50 to 200 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
F123615GPR 20 to 100 μL
F144801GPR 0.2 to 2 μL
F144802GPR 1 to 10 μL
F123600GPR 2 to 20 μL
F123601GPR 50 to 200 μL
F123602GPR 200 to 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. F123615GPR Supplier Gilson™ Supplier No. F123615PR
Gilson™ PIPETMAN Classic™ Pipets are individually calibrated, delivered with a performance check report, and include dry seal technology for air-tightness.
  • Fully adjustable, air-displacement pipette with selected volume shown on digital indicator (volumeter)
  • Tip holder and tip ejector are autoclavable to avoid cross-contamination
  • Each pipet bears engraved identification number for traceability
Specifications
Accuracy ±0.35, ±1.8 μL
Compatible Tips D200, DF100ST
Material PVDF, Stainless Steel
Model P100
Product Line Pipetman Classic
Type Pipette
Volume (Metric) 20 to 100 μL
For Use With (Application) Small volume dispensing of aqueous fluids of moderate viscosity and density (molecular biology, immunology)