Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMANTM Diamond Blister Pack Refill Pipette Tips
GREENER_CHOICE

Catalog No. F172310G Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
100 to 1000 μL
2 to 200 μL
2 to 30 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility
F172310G 2 to 200 μL Non-filtered Non-sterile
F172511G 100 to 1000 μL Non-filtered Sterile
F172510G 100 to 1000 μL Non-filtered Non-sterile
F172313G 2 to 30 μL Filtered Sterile
F172311G 2 to 200 μL Non-filtered Sterile
F172613G 100 to 1000 μL Filtered Sterile
F172513G 2 to 200 μL Filtered Sterile
F172213G 0.1 to 10 μL Filtered Sterile
F172210G 0.1 to 20 μL Non-filtered Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. F172310G Supplier Gilson™ Supplier No. F172310
Add to Cart
Add to Cart

Autoclavable or gamma-ray sterlized

  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Standard or filtered tips
  • Validated for use with 384-well tips: F172211G, F172311G and F17511G

Specifications

Volume (Metric) 2 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Model DS200
Length (English) 10.79 in.
Length (Metric) 51 mm
Clean Claims Manufactured in clean room
For Use With (Application) Pipetting
Sterilization Method Autoclavable
Format Blister Refill
Tip Style Universal
Product Type Air Displacement Tips
Color Code Yellow
For Use With (Equipment) PIPETMAN Pipettes
Product Line Pipetman Diamond
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.