Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMANTM Diamond Blister Pack Refill Pipette Tips
GREENER_CHOICE

Catalog No. F172613G Shop All Gilson Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
0.1 to 20 μL
100 to 1000 μL
2 to 200 μL
2 to 30 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Filtered Sterility
F172613G 100 to 1000 μL Filtered Sterile
F172511G 100 to 1000 μL Non-filtered Sterile
F172510G 100 to 1000 μL Non-filtered Non-sterile
F172313G 2 to 30 μL Filtered Sterile
F172311G 2 to 200 μL Non-filtered Sterile
F172513G 2 to 200 μL Filtered Sterile
F172310G 2 to 200 μL Non-filtered Non-sterile
F172213G 0.1 to 10 μL Filtered Sterile
F172210G 0.1 to 20 μL Non-filtered Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. F172613G Supplier Gilson™ Supplier No. F172613
Add to Cart
Add to Cart

Autoclavable or gamma-ray sterlized

  • Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
  • Features graduations at varying volumes allowing for quick-checks when aspirating samples
  • Standard or filtered tips
  • Validated for use with 384-well tips: F172211G, F172311G and F17511G

Specifications

Volume (Metric) 100 to 1000 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Model DSF1000
Length (English) 10.79 in.
Length (Metric) 66 mm
Clean Claims DNA free, RNA free, RNAse free, DNAse free, ATP free, Endotoxin free
For Use With (Application) Pipetting
Sterilization Method Gamma radiation
Format Refill
Tip Style Straight
Product Type Air Displacement Tips
Color Code Blue
For Use With (Equipment) Gilson Pipetmax
Product Line Pipetman Diamond
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.