Learn More
United Scientific™ Pipette Filling Device Stand
SDPSCI_ED
Catalog No. S43743 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Height (Metric):
12.5 cm
Catalog No. Height (Metric)
S43743 12.5 cm
1 options
Catalog No. S43743 Supplier United Scientific™ Supplier No. PFDS01
Safely store pipette filling devices with this Pipette Filling Device Stand.
  • For use with United Scientific Pipette Filling Device (PFD001).
  • Compatible with many other brands.
Specifications
Material Polycarbonate
Height (Metric) 12.5 cm
Product Type Device Stand
For Use With (Equipment) Pipette Filling Device (PFD001)
Quantity 1 Ea.