Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Pipette Pumps
SDPSCI_ED

Catalog No. S00640 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Volume (Metric):
2 mL
10 mL
25 mL
Color:
Blue
Green
Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
S00640 2 mL Blue
S00641 10 mL Green
S00642 25 mL Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S00640 Supplier United Scientific™ Supplier No. PPMP02PK/12
Add to Cart
Add to Cart
This item is not returnable. View return policy

Pipette Pumps work with a wide variety of pipettors and allow simple, one handed use.

  • Good quality pipettors designed for easy and precise pipetting.
  • Pumps feature a lever for quick release, and forward or backward wheel movement.
  • The soft elastic chuck has a threaded collar to tightly grip glass or plastic pipettes.
  • Easy to clean.

Specifications

Adjustable Tip Spacing No
Autoclavable Not Autoclavable
Color Blue
Disposable Reusable
For Use With Glass or Plastic Pipettes
Grip Type Ergonomic
Material Plastic
Model PPMP02
Number of Channels 1
Sterility Non-sterile
Type Pipette Pump
Volume (Metric) 2 mL
Quantity 12 Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.