Learn More
Andwin Scientific 4-Place Pipet Racks Acrylic pipet racks
SDP
Supplier:  Andwin Scientific 82024536
Encompass_Preferred
4-Place Pipet Racks Acrylic pipet racks keep instruments secure and conveniently accessible in the laboratory environment. Accommodate 0.1 to 25mL pipets or cans. Clear plastic construction. Feature four angled compartments. Dimensions: 8.9x29.2x40.6c
Catalog No. NC9860108
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.