Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Pipette Tip Receptacle
SDP

Catalog No. 22000759
Change view
Click to view available options
Includes:
Magnet Mount
VHB Tape
Wall Mount
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-000-759 VHB Tape
22-000-758 Magnet Mount
22-000-760 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-000-759 Supplier Poltex Supplier No. TIPDISPT
Add to Cart
Add to Cart
This item is not returnable. View return policy

Receptacle for used pipette tips.

  • Back plate mounts to wall, inside of hood, etc.
  • Front receptacle lifts off for easy emptying/cleaning.
  • Choose from VHB tape mount (very high bond tape included), or magnet mount (powerful magnets included).

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Pipettes
Material PETG
Positions 1
Shape Rectangular, Rounded Corners
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipette
Length (Metric) 11 cm
Includes VHB Tape
Product Type Pipet Rack
Array 1 x 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.