Learn More
Mascon Medical Eppendorf 1250L Pipette Tips, manual, sterile, filtered,PCR, 50 Rack/Case
SDP
Supplier:  Mascon Medical 800515
Encompass_Preferred
Mascon Medical™ offers an extensive line of compatible robotic tips for Hamilton, Eppendorf™, Rainin and Beckman automation systems. These high precision tips are manufactured to guarantee a perfect fit and extremely accurate dispensing behavior. Available in non-filtered and filtered. Nonpyrogenic. Conductive Polypropylene tips are available in 96-well nested tip rack.Features: 96 Tip/Nested Tip Rack Format Nonpyrogenic Sterile Filtered Specifications: Volume: 1250L Qty. / Case: 4800 Tip Shape: Round Tip Format: 96 Tips/Rack Material: Polypropylene Color: Transparent Filter: Yes Sterile: Yes Packaging: Nested Tip Rack, 10r Racks/Pack, 5 Packs /Carton Packaging Format: 50 Racks/Carton
Catalog No. NC2180265
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.