Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Pipetter Nozzles
SDP

Catalog No. 22146656 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
100 to 200 μL
2 μL
2 to 20 μL
5 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-146-656 2 μL
22-146-660 100 to 200 μL
22-146-653 2 to 20 μL
22-146-244 0.5 to 20 μL
22-146-306 5 mL
22-146-664 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-146-656 Supplier VistaLab™ Technologies Supplier No. 90571013
Add to Cart
Add to Cart

Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter

  • Available in range of sizes

Specifications

For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 2 μL
For Use With (Equipment) Adjustable Models 0.2 μL, Fixed Models 1 μL and 2 μL
Includes Filter
Quantity 5/Pk.
Length (English) 2 in.
Width (English) 1 in.
Height (English) 1 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.