Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VistaLab™ Technologies Pipetter Nozzles
SDP

Catalog No. 22146664 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
100 to 200 μL
2 μL
2 to 20 μL
5 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
22-146-664 -
22-146-660 100 to 200 μL
22-146-653 2 to 20 μL
22-146-656 2 μL
22-146-244 0.5 to 20 μL
22-146-306 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 22-146-664 Supplier VistaLab™ Technologies Supplier No. 90573010

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter

  • Available in range of sizes

Specifications

For Use With Pipettes
Material Plastic
For Use With (Equipment) 25 to 1250 μL Electronic Single Channel Model, 100 to 1000 μL and 20 to 200 μL Adjustable Model, 201 to 1000 μL Fixed Models
Includes Internal aerosol/liquid barrier filter
Quantity 10/Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.