Learn More
VistaLab™ Technologies Pipetter Power Supplies and Batteries
SDP
Catalog No. 22147109 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Type:
Lithium-ion Battery
Power Supply
Voltage:
3.7 V
100 V
230 V
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Voltage
22-147-109 Power Supply 100 V
22-147-112 Lithium-ion Battery 3.7 V
22-147-110 Power Supply 230 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 22-147-109 Supplier VistaLab™ Technologies Supplier No. 90609002
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Specifications
Battery Type Lithium-ion
For Use With Pipettes
Material Plastic
Type Power Supply
For Use With (Equipment) Ovation™ Electronic Single Channel, Macro and Multichannel Models
Electrical Requirements 100 V, 50/60 Hz
Voltage 100 V
Frequency 50/60 Hz