Learn More
TARGETMOL CHEMICALS INC PIPROZOLIN 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T34076
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Piprozolin (W 3699) is a new type of choleretic acid that shows slight analgesic properties at high doses during chronic experiments.Piprozolin has a slight induction of certain enzymes of the oxidative system. purity: 99%
Catalog No. 50-258-0280
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.