Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Piracetam, AMPA Receptor Positive Allosteric Modulator, Solid, 50 mg

Catalog No. 502706659
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6659 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6659 Supplier Abcam Supplier No. AB14138050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nootropic agent that acts as a positive allosteric modulator at AMPA receptors.

  • Molecular Weight: 142.16 Da
  • Form: Solid
  • CAS Number: 7491-74-9
  • Mechanism of Action: Positive allosteric modulator at AMPA receptors
  • Description: Nootropic agent and GABA derivative that potentiates cholinergic and glutamatergic neurotransmission via modulation of ion flux
  • Biological Effects: Includes neuroprotection, anticonvulsant properties, and reduced vascular endothelium adhesion
  • Activity: Active in vivo
  • Composition: 100% Piracetam

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule AMPA Receptors
Form Solid
Buffer 100% Piracetam
Content And Storage Store at ambient temperature.
Formulation 100% Piracetam
Quantity 50 mg
For Use With (Application) Neuroscience research, neuroprotection, anticonvulsant studies
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.