Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pirenzepine dihydrochloride, R&D Systems™
SDP

Catalog No. 1071100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-711-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-711-00 Supplier R&D Systems Supplier No. 1071/100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective M1 antagonist

M1 muscarinic receptor selective antagonist. Inverse agonist activity reported.

Chemical Identifiers

CAS 29868-97-1
Molecular Formula C19H23Cl2N5O2
Molecular Weight (g/mol) 424.326
InChI Key FFNMBRCFFADNAO-UHFFFAOYSA-N
Synonym pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
PubChem CID 71405
ChEBI CHEBI:32014
IUPAC Name 11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
SMILES CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl

Specifications

Quantity 100 mg
Formula Weight 424.33
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Pirenzepine dihydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.