Learn More
Clontech Labs 3P PIRES VECTOR
SDP
Supplier:  Clontech Labs 3P 631605
Encompass
pIRES Vector 20microgram Uses: Permits two genes of interest to be translated from a single RNA Type: bicistronic expression Promoter:cmV IE
Catalog No. NC9658061
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.