Learn More
Clontech Labs 3P PIRES2-DSRED2 VECTOR 20UG
SDP
Supplier:  Clontech Labs 3P 632420
Encompass
Bicistronic expression vector containing DsRed2 as a marker for transfection efficiency. A gene of interest is inserted into the MCS located upstream of the encephalomyocarditis virus EMCV internal ribosomal entry site IRES. For Reasearch Use ONLY.
Catalog No. NC2383936
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.