Learn More
Apexbio Technology LLC Piriprost (potassium salt) 88851-62-1 50mg
SDP
Supplier:  Apexbio Technology LLC C482850
Encompass_Preferred
Piriprost (potassium salt CAS 88851-62-1) also known as U 60257 is a small molecule inhibitor of leukotriene biosynthesis Leukotrienes are inflammatory mediators derived from eicosanoid metabolism in leukocytes and act via G protein-coupled receptors By inhibiting 5-lipoxygenase-mediated production of leukotriene B4 sulfidopeptide leukotrienes and the metabolite 5-HETE Piriprost reduces leukotriene-driven inflammatory processes in experimental mast cell and basophil leukemia models exhibiting IC50 values in the 9 50 M range Piriprost has also been investigated for effects on pulmonary permeability endometrial cell alkaline phosphatase activity and alveolar epithelial integrity offering utility in inflammatory and respiratory research
Catalog No. 50-000-63393
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.