Learn More
StatLab™ PiSmart Thermal Transfer Microbiopsy Cassettes
Description
- 1,000 per box, with 2 boxes per case for a total of 2,000 per case.
- Threaded in strips of 40 with lids separate.
- PiSmart cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing.
- Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer.
- Available in 9 colors.
Specifications
Specifications
| Color | Blue |
| Product Type | Microbiopsy Cassette |