Learn More
3M™ Pistachio Protein Rapid Kit L25PST, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products

Description
- Results visible at 11 ± 1 min. after application of sample
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for detection of allergenic food proteins
- Designed to detect both processed and unprocessed allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Specifications
| Format | Kit |
| Product Type | Pistachio Protein Rapid Kit |
| Test Type | Allergen |
| For Use With (Application) | Screening pistachio proteins in CIP final rinse water, environmental swab samples, food ingredients and processed food product |
| Quantity | 25/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.