Learn More
Apexbio Technology LLC AZD8931 (Sapitinib) 848942-61-0 50mg
SDP
Supplier:  Apexbio Technology LLC A837550
Encompass_Preferred
AZD8931 (Sapitinib CAS 848942-61-0) is a reversible small-molecule inhibitor that targets EGFR (HER1) HER2 (erbB2) and HER3 (erbB3) with similar potency It suppresses receptor phosphorylation with IC50 values of 4 nM (EGFR) 3 nM (HER2) and 4 nM (HER3) Preclinical studies using HER2-non-amplified inflammatory breast cancer (IBC) cell lines demonstrate reduced tumor cell proliferation and increased apoptosis upon AZD8931 administration In mouse xenograft models AZD8931 alone or combined with paclitaxel significantly delays tumor growth AZD8931 is under clinical evaluation as combination therapy in advanced breast cancer
Catalog No. 50-000-61720
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.