Learn More
Invitrogen™ PITPN Polyclonal Antibody

Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
Specifications
Specifications
| Antigen | PITPN |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PITPNA |
| Gene Accession No. | Q00169 |
| Gene Alias | epididymis secretory protein Li 36; HEL-S-36; phosphatidylinositol transfer protein alpha; phosphatidylinositol transfer protein alpha isoform; phosphatidylinositol transfer protein, alpha; Pitp alpha; PI-TPalpha; PI-TP-alpha; PITPN; Pitpna; PITPNB; ptdIns transfer protein alpha; ptdInsTP alpha; vb; VIB1A; vibrator |
| Gene Symbols | PITPNA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.