Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITPNB Monoclonal Antibody (OTI5D11), TrueMAB™, OriGene
SDP

Catalog No. 501682737
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2737 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2737 Supplier OriGene Supplier No. TA809354
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The lipid binding proteins known as phosphatidylinositol transfer proteins (PITP) facilitate the formation of phosphatidylinositol derived second messenger molecules, which are related to the phospholipase C and phosphoinositide 3-kinase pathways. PITP are ubiquitously expressed proteins that transfer phosphatidylinositol (PI) and phosphatidylcholine (PC) between membranes enriched in PI or PC to membranes that are deficient in PI or PC. PITP mobilizes PI from the endoplasmic recticulum and regulates the release of PI from stored vesicles in the Golgi network. In mammalian cells, three smaller forms of soluble PITP are present, designated PITP alpha, PITP beta and retinal degeneration B (rdgB) beta. PITP beta is a 271 amino acid protein that is widely expressed in various tissues. Though required for Golgi targeting, constitutive phosphorylation of Ser-262 has no effect on phospholipid transfer activity. There are two isoforms of PITP beta that are produced as a result of alternative splicing events.

Specifications

Antigen PITPNB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PITPNB
Gene Accession No. P48739
Gene Alias PI-TP-beta, PtdInsTP, VIB1B
Gene Symbols PITPNB
Host Species Mouse
Immunogen Full length human recombit protein of human PITPNB produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23760
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.