Learn More
Invitrogen™ PITPNB Polyclonal Antibody

Description
Recommended positive controls: 293T, A431 , Jurkat, Raji. Predicted reactivity: Mouse (97%), Rat (98%), Zebrafish (85%), Chicken (92%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | PITPNB |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | PITPNB |
| Gene Accession No. | P48739 |
| Gene Alias | phosphatidylinositol transfer protein beta; phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; Pitp beta; PI-TP beta; PI-TP-beta; Pitpnb; ptdIns transfer protein beta; PtdInsTP; ptdInsTP beta; VIB1B |
| Gene Symbols | PITPNB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.