Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Pitstop 2-100, Clathrin Terminal Domain Inhibitor, Solid, 5 mg
Description
- Chemical class: Clathrin terminal domain inhibitor
- Purity: >99%
- Molecular weight: 463.35 g/mol
- Potency: IC50 = 7.5 μM
- Key biological activity: Specifically inhibits the internalisation of major histocompatibility complex 1 (MHC1)
- Physical form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Clathrin |
| Buffer | 100% Pitstop™ 2-100 |
| Form | Solid |
| Content And Storage | Store at 4°C. |
| Formulation | 100% Pitstop™ 2-100 |
| Quantity | 5 mg |
| For Use With (Application) | Clathrin terminal domain inhibition, MHC1 internalisation inhibition |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?