Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Pitstop™ 2-100 Negative Control, Solid, 5 mg
Description
- Molecular weight: 463.39 g/mol
- Form: Solid
- Function: Negative control for clathrin inhibitor Pitstop™ 2-100
- Performance: Structurally related to Pitstop™ 2-100; no effect on clathrin-mediated endocytosis of transferrin or major histocompatibility complex 1 uptake
Specifications
Specifications
| Target Molecule | Clathrin |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% Pitstop™ 2-100 - negative control |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Formulation | 100% Pitstop™ 2-100 - negative control |
| Quantity | 5 mg |
| For Use With (Application) | Negative control for clathrin inhibitor Pitstop 2-100 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?