Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552600
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552600 Supplier Invitrogen™ Supplier No. PA552600
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFTFFNSMSP LSSQSMFSAP SSISSMTMPS SMGPGAVPGM PNSGLNNINN LTGSSLNSAM SPGACPYGTP ASPYSVYRDT CNSSLASLRL KSKQHSS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 99%.

PITX1 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family are involved in organ development and left-right asymmetry. This protein acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITX1
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PITX1
Gene Accession No. P78337
Gene Alias BFT; CCF; hindlimb expressed homeobox protein backfoot; hindlimb-expressed homeobox protein backfoot; homeobox protein PITX1; Homeobox protein P-OTX; LBNBG; OTX; paired like homeodomain 1; paired-like homeodomain 1; paired-like homeodomain transcription factor 1; pituitary homeo box 1; pituitary homeobox 1; Pituitary OTX-related factor; Pitx1; Potx; P-OTX; PTX1; Unknown (protein for MGC:136976); zgc:136976
Gene Symbols PITX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PITX1. Recombinant protein control fragment (Product #RP-89293).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5307
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.