Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PI382850 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI382850 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI382850 Supplier Invitrogen™ Supplier No. 382850
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PITX3 Polyclonal Antibody for IHC (F), ICC/IF

This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family act as transcription factors. This protein is involved in lens formation during eye development. Mutations of this gene have been associated with anterior segment mesenchymal dysgenesis and congenital cataracts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITX3
Applications Immunohistochemistry (Frozen), Immunocytochemistry, Transcription Factor Activity and Detection
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Dilution Assay-dependent
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PITX3
Gene Accession No. O35160
Gene Alias ak; aphakia; ASMD; ASOD; CTPP4; CTRCT11; Homeobox protein PITX3; homeobox protein PTX3; MGC12766; paired like homeodomain 3; paired-like homeodomain 3; paired-like homeodomain transcription factor 3; Pituitary homeobox 3; Pituitary homeobox 3 (Homeobox protein PITX3); PITX3; Ptx3
Gene Symbols PITX3
Host Species Rabbit
Immunogen Synthetic peptide derived from the N-terminal region of the mouse PiTX3 protein.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Research Discipline Neurobiology
Primary or Secondary Primary
Gene ID (Entrez) 18742
Packaging 1 Each
Includes 0.1% Sodium azide
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.