Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pivalaldehyde 95.0+%, TCI America™
SDP

Catalog No. P084725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P084725ML 25 mL
P08475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P084725ML Supplier TCI America Supplier No. P084725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 630-19-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006962
InChI Key FJJYHTVHBVXEEQ-UHFFFAOYSA-N
Synonym trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
PubChem CID 12417
IUPAC Name 2,2-dimethylpropanal
SMILES CC(C)(C)C=O

Specifications

Melting Point 6°C
Color Yellow
Boiling Point 75°C
Quantity 25 mL
UN Number 1989
Formula Weight 86.13
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Pivalaldehyde
EINECSNumber (2)-0494
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.