Learn More
CORRELIA BIOSYSTEMS PIXI CARTRIDGE
Supplier:  CORRELIA BIOSYSTEMS 377IGGG
Encompass
For PIXI system. cartridge combines a nonporous capture substrate with electrically-driven mass transport to eliminate the slow diffusive steps which limit the efficiency of traditional immunoassays
Catalog No. NC2302693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.