Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PJ 34 hydrochloride
GREENER_CHOICE

Catalog No. p-4784305
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-551-0 10 mg
32-555-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 32-551-0 Supplier Tocris Bioscience™ Supplier No. 3255/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent PARP inhibitor

Potent inhibitor of poly(ADP-ribose) polymerase (PARP) (EC50 = 20 nM). ∽1000-fold more potent than 3-Aminobenzamide (Cat. No. 0788). Protects primary neuronal cells from oxygen-glucose deprivation in vitro and reduces infarct size following focal cerebral ischemia in vivo. Displays protective effects against cisplatin -induced kidney injury. Also displays activity at Pim-1 and Pim-2 kinases at higher concentrations (IC50 values are 3.7 and 16 μM respectively).

Chemical Identifiers

CAS 344458-15-7
Molecular Formula C17H18ClN3O2

Specifications

Quantity 10 mg
Formula Weight 331.8
Purity >98%
Molecular Weight (g/mol) 331.8
Chemical Name or Material PJ 34 hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.