Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PJA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559142
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559142 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559142 Supplier Invitrogen™ Supplier No. PA559142
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GIALVHTDSY DPDGKHGEDN DHLQLSAEVV EGSRYQESLG NTVFELENRE AEAYTGLSPP VPSFNCEVRD EFEELDSVPL VKSSAGDTEF VH Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 64%.

Has E2-dependent E3 ubiquitin-protein ligase activity. Responsible for ubiquitination of cAMP-dependent protein kinase type I and type II-alpha/beta regulatory subunits and for targeting them for proteasomal degradation. Essential for PKA-mediated long-term memory processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PJA2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PJA2
Gene Accession No. O43164
Gene Alias AI447901; AL022700; E3 ubiquitin-protein ligase Praja2; E3 ubiquitin-protein ligase Praja-2; Kiaa0438; mKIAA0438; Neurodap1; neurodegeneration associated protein 1; Pja2; praja 2, RING-H2 motif containing; praja ring finger 2, E3 ubiquitin protein ligase; praja ring finger ubiquitin ligase 2; praja2; praja-2; protein carrying the RING-H2 sequence motif; RING finger protein 131; RING-type E3 ubiquitin transferase Praja-2; RNF131
Gene Symbols PJA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PJA2. Recombinant protein control fragment (Product #RP-96982).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9867
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.