Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKC alpha/beta/gamma Recombinant Rabbit Monoclonal Antibody (9O5J1)

Catalog No. MA555525
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA555525 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA555525 Supplier Invitrogen™ Supplier No. MA555525
  • $474.00
    $355.65 / Each of 1

    Save $118.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: ELRQKFERAK ISQGTKVPEE KTTNTVSKFD NNGNRDRMKL TDFNFLMVLG KGSFGKVMLS ERKGTDELYA VKILKKDVVI QDDDVECTMV EKRVLALPGK PPFLTQLHSC FQTMDRLYFV MEYVNGGDLM YHIQQVGRFK EPHAVFYAAE IAIGLFFLQS KGIIYRDLKL DNVMLDSEGH IKIADFGMCK ENIWDGVTTK T.

Protein Kinase c alpha (PKC alpha) is an 77 kDa member of the conventional group (cPKCs: sensitive to calcium, diacylglycerol, phosphatidylserine and phorbol esters) of the PKC family of serine/ threonine kinases that are involved in a wide range of physiological processes including mitogenesis, cell survival and transcriptional regulation. PKC alpha is an ubiquitously expressed PKC isozyme that has been implicated in the regulation of a broad range of cellular functions including proliferation, differentiation, development, migration, cell cell adhesion, cell extracellular matrix adhesion, and solute transport. The activation loop threonine (threonine 497 in PKC alpha) of conventional PKCs is phosphorylated by phosphoinositide dependent kinase 1 (PDK1). This phosphorylation is necessary for the autophosphorylation of threonine 638 in the carboxy terminus of PKC alpha, a step that is critical for regulating the rate of PKC alpha dephosphorylation and inactivation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC alpha/beta/gamma
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9O5J1
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene PRKCG
Gene Accession No. P05129, P05696, P05771, P17252, P20444, P63318, P63319, P68403, P68404
Gene Alias A130082F03Rik; cAMP-dependent protein kinase catalytic subunit alpha; cAMP-dependent protein kinase catalytic subunit alpha, isoform 1; cAMP-dependent protein kinase catalytic subunit C alpha; Cs-PKA; PKA C-alpha; Pkaca; PKC; PKCA1; Pkcb; PKC-B; PKC-beta; PKCC; PKCD; PKCG; PKCgamma; PKC-gamma; PKCI; PPNAD4; PRKACA; Prkc; Prkcb; Prkcb1; Prkcb2; Prkcc; Prkcg; protein kinase A; protein kinase A alpha; protein kinase A catalytic subunit; protein kinase C beta; protein kinase C beta I; protein kinase C beta II; protein kinase C beta type; protein kinase C beta-II; protein kinase C gamma; protein kinase C gamma type; protein kinase C type I (gamma type); protein kinase C, beta; protein kinase C, beta 1; protein kinase C, beta 1 polypeptide; protein kinase C, gamma; protein kinase cAMP-activated catalytic subunit alpha; protein kinase, cAMP dependent, catalytic, alpha; protein kinase, cAMP-dependent, alpha catalytic subunit; protein kinase, cAMP-dependent, catalytic, alpha; RATPKCI; SCA14; sperm cAMP-dependent protein kinase catalytic subunit
Gene Symbols PRKCA, PRKCB, PRKCG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 300-500 of human PKC alpha + beta + gamma.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18750, 18751, 18752, 24680, 24681, 25023, 5578, 5579, 5582
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.