Learn More
Invitrogen™ PKC lambda/iota Polyclonal Antibody

Description
Immunogen sequence: STMSHTVAGGG SGDHSHQVRV KAYYRGDIMI THFEPSISFE GLCNEVRDMC SFDNEQLFTM KWIDEEGDPC TVSSQLEL Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.
Specifications
Specifications
| Antigen | PKC lambda/iota |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PRKCI |
| Gene Accession No. | P41743 |
| Gene Alias | 2310021H13Rik; AI427505; aPKClambda; aPKC-lambda/iota; atypical protein kinase C-lambda/iota; DXS1179E; hypothetical protein; mKIAA4165; nPKC-iota; Pkci; Pkcl; PKClambda; PRKCI; Prkcl; PRKC-lambda/iota; protein kinase C iota; protein kinase C iota type; protein kinase C, iota; protein kinase C, lambda |
| Gene Symbols | PRKCI |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.