Learn More
Abcam PKC Pathway Modulator Set, Chemical Compounds, 1 KIT
Description
- Set of PKC pathway modulators
- Target proteins include PKC, PKC alpha, PKC delta, JAK1, Axl, JAK3, VEGF Receptor 2, Fyn, GSK3 beta, Cytochrome P450 3A4, and CDK2
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | PKC, PKC Alpha, PKC Delta, JAK1, Axl, JAK3, VEGF Receptor 2, Fyn, GSK3 Beta, Cytochrome P450 3A4, CDK2 |
| Format | Kit |
| Product Type | Western Blot Kit |
| Quantity | 1 Kit |
| For Use With (Application) | PKC pathway modulation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.