Learn More
Description
This antibody detects a 79 kDa* protein corresponding to the molecular mass of PKCtheta on SDS-PAGE immunoblots of human Jurkat cell lysates.
Specifications
Specifications
| Antigen | PKC theta |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | M217 |
| Concentration | 0.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | PRKCQ |
| Gene Accession No. | Q02111, Q04759, Q9WTQ0 |
| Gene Alias | A130035A12Rik; AW494342; EC 2.7.11.13; kinase nPKC-theta; KPCT; MGC126514; MGC141919; nPKC-theta; PKC theta; PKC-0; Pkcq; PKCtheta; PKC-theta; PRKCQ; PRKCT; protein kinase C theta; protein kinase C theta type; protein kinase C, theta; RP11-563J2.1; si:ch211-239o15.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
