Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKC theta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596442
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596442 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596442 Supplier Invitrogen™ Supplier No. PA596442
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSPFLRIGLS NFDCGSCQSC QGEAVNPYCA VLVKEYVESE NGQMYIQKKP TMYPPWDSTF DAHINKGRVM QIIVKGKNVD LISETTVELY SLAERCRKNN GKTEIWLELK PQGRMLMNAR YFLEMSDTKD MNEFETEGFF ALHQRRGAIK QAKVHHVKCH EFTATFFPQP TFCSVCHEFV WGLNKQGYQC RQCNAAIHKK CIDKVIAKCT GSAINSRETM FHKERFKIDM; Positive Samples: HepG2, Rat testis; Cellular Location: Cell membrane, Cytoplasm, Peripheral membrane protein.

Protein Kinase C (PKC) isoforms are serine/threonine kinases involved in signal transduction pathways that govern a wide range of physiological processes including differentiation, proliferation, gene expression, brain function, membrane transport and the organization of cytoskeletal and extracellular matrix proteins. Increasing evidence from studies using in vitro and in vivo systems points to PKC as a key regulator of critical cell cycle transitions, including cell cycle entry and exit and the G1 and G2 checkpoints. PKC-mediated control of these transitions can be negative or positive, depending on the timing of PKC activation during the cell cycle and on the specific PKC isozymes involve. There have been at least 12 different PKC isoforms identified in humans to date, including alpha, beta I, beta II, gamma, delta, epsilon, zeta, eta, theta, iota, lambda, and mu. The different PKC isoforms vary in structure, subcellular localization, tissue distribution and response to extracellular signals. PKC beta II has been shown to be expressed, in part, in cardiac muscle, intestinal epithelium, kidney and pancreas. PKC beta II has been shown to be involved in cell cycle progression and cancer proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKC theta
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PRKCQ
Gene Accession No. Q02111, Q04759, Q9WTQ0
Gene Alias A130035A12Rik; AW494342; EC 2.7.11.13; kinase nPKC-theta; KPCT; MGC126514; MGC141919; nPKC-theta; PKC theta; PKC-0; Pkcq; PKCtheta; PKC-theta; PRKCQ; PRKCT; protein kinase C theta; protein kinase C theta type; protein kinase C, theta; RP11-563J2.1; si:ch211-239o15.1
Gene Symbols PRKCQ
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human PRKCQ (NP_0062481).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18761, 5588, 85420
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.