Learn More
Invitrogen™ Phospho-PKC zeta (Thr560) Recombinant Rabbit Monoclonal Antibody (8Z5E6)

Description
Immunogen sequence: QLTPD.
Specifications
Specifications
| Antigen | Phospho-PKC zeta (Thr560) |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 8Z5E6 |
| Concentration | 1.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PRKCZ |
| Gene Accession No. | P09217, Q05513 |
| Gene Alias | 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; AI098070; aPKCzeta; atypical protein kinase C; C80388; kinase nPKC-zeta; KPCZ; nPKC-zeta; nPKC-zeta PRKCZ; PKC2; Pkcz; PKC-ZETA; Prkcz; protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, zeta; r14-3-3; R74924; zetaPKC |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.