Learn More
Invitrogen™ PKI [14 22] Myristoylated PTD
Description
cAMP-dependent protein kinase (PKA) inhibitor. This peptide is a highly specific inhibitor of cAMP-dependent protein kinase (PKA).
Format/Formulation: Lyophilized trifluoroacetate salt, >97% purity by HPLC.
Molecular Weight: 1208
Specifications
Specifications
| Content And Storage | Store in freezer (-5 to -30°C). |
| Gene ID (Entrez) | 5566, 5567, 5568 |
| Purification Method | HPLC |
| Shipping Condition | Dry Ice |
| Specific Inhibit or Target | PKA |
| Sterility | Non-sterile |
| Quantity | 1 mg |
| Product Type | cAMP-Dependent Protein Kinase (PKA) Inhibitor |
| Inhibitor Properties | Protein |
| Molecular Weight (g/mol) | 1209 |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.