Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM1 Rabbit anti-Human, Mouse, Rat, Biotin, Polyclonal, Proteintech
SDP

Catalog No. 501737222 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7222 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7222 Supplier Proteintech Group Inc Supplier No. BIOTIN15821100UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody is conjugated with Biotin and it's specific to PKM1 isoform.

PKM2 is a pyruvate kinase that catalyzes the production of phosphoenolpyruvate from pyruvate and ATP. PKM2 has been shown to interact with thyroid hormone, and thus may mediate cellular metabolic effects induced by thyroid hormones. PKM2 has been found to bind Opa protein, a bacterial outer membrane protein involved in gonococcal adherence to and invasion of human cells, suggesting a role of this protein in bacterial pathogenesis. Three alternatively spliced transcript variants encoding two distinct isoforms of PKM2 have been reported.

Specifications

Antigen PKM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and no preservative; pH 7.3
Gene PKM
Gene Accession No. P11980, P14618, P52480
Gene Alias CTHBP, OIP 3, OIP3, Opa interacting protein 3, p58, PK2, PK3, PKM, PKM1, PKM2, PKM2 pyruvate kinase, muscle, Pyruvate kinase 2/3, Pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, TCB, THBP1, Tumor M2 PK
Gene Symbols Pkm
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18746, 25630, 5315
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.