Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PKN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572912
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572912 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572912 Supplier Invitrogen™ Supplier No. PA572912
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKN1 Polyclonal Antibody for Western Blot, IHC (P), IP

PKN1 (PRK1) belongs to the protein kinase C superfamily. This kinase is activated by the Rho family of small G proteins and may mediate the Rho-dependent signaling pathway. This kinase can also be activated by phospholipids. The 3-phosphoinositide dependent protein kinase-1 (PDPK1/PDK1) is reported to phosphorylate this kinase, which may mediate insulin signals to the actin cytoskeleton. The proteolytic activation of this kinase by caspase-3 or related proteases during apoptosis suggests its role in signal transduction related to apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PKN1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 0.05% sodium azide
Gene PKN1
Gene Accession No. P70268, Q16512, Q63433
Gene Alias DBK; F730027O18Rik; PAK1; PAK-1; PKN; PKN1; PKN-ALPHA; PRK1; Prkcl1; Protease-activated kinase 1; protein kinase C-like 1; Protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase N1; protein kinase PKN; protein kinase PKN-alpha; Protein-kinase C-related kinase 1; serine/threonine kinase 3; serine/threonine protein kinase N; serine/threonine-protein kinase N1; serine-threonine kinase N; serine-threonine protein kinase N; Stk3
Gene Symbols PKN1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 911-929 of human PKN1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100856398, 29355, 320795, 5585
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.