Learn More
Medchemexpress LLC PKUMDL-LC-101-D04 25mg | 2143896-83-5 | 378.9 | C14H22N4O2S2 • HCl | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11562725MG
Encompass_Preferred
PKUMDL-LC-101-D04 (GPX4-Activator-1d4) is a small-molecule allosteric activator of glutathione peroxidase 4 (GPX4). It increases GPX4 enzymatic activity (pEC50 = 4.7) in biochemical and cell-based assays and has been reported to inhibit ferroptosis and inflammatory responses. The compound is supplied as the monohydrochloride salt with reported high purity and is intended for research use.
Catalog No. 50-002-28520
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.