Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA2G2D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548132
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548132 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548132 Supplier Invitrogen™ Supplier No. PA548132
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLA2G2D Polyclonal Antibody for Western Blot, IHC (P), Flow

PA2 catalyzes the calcium-dependent hydrolysis of the 2- acyl groups in 3-sn-phosphoglycerides. L-alpha-1-palmitoyl-2- linoleoyl phosphatidylethanolamine is more efficiently hydrolyzed than the other phospholipids examined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G2D
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Pla2g2d
Gene Accession No. Q9UNK4
Gene Alias GIID sPLA2; Group IID secretory phospholipase A2; mGIID; phosphatidylcholine 2-acylhydrolase 2D; phospholipase A2 group IID; phospholipase A2, group IID; Pla2a2; PLA2G2D; PLA2IID; secretory phospholipase A2s; secretory-type PLA, stroma-associated homolog; sPLA2-IID; sPLA2S; SPLASH
Gene Symbols Pla2g2d
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 105-131 amino acids from the C-terminal region of human PLA2G2D.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26279
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.