Learn More
Invitrogen™ PLA2G2D Polyclonal Antibody

Description
Immunogen sequence: GILNLNKMVK QVTGKMPILS YWPYGCHCGL GGRGQPKDAT DWCCQTHDCC YDHLKTQGCS IYKDYYRYNF SQGNIHCSDK GSWCEQQLCA CDKEVAFCLK RNLDTYQKRL RFYWRPHCRG QTPGC; Positive Samples: HL-60, THP-1.
Specifications
Specifications
| Antigen | PLA2G2D |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.78 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 |
| Gene | Pla2g2d |
| Gene Accession No. | Q9UNK4, Q9WVF6 |
| Gene Alias | GIID sPLA2; Group IID secretory phospholipase A2; mGIID; phosphatidylcholine 2-acylhydrolase 2D; phospholipase A2 group IID; phospholipase A2, group IID; Pla2a2; PLA2G2D; PLA2IID; secretory phospholipase A2s; secretory-type PLA, stroma-associated homolog; sPLA2-IID; sPLA2S; SPLASH |
| Gene Symbols | Pla2g2d |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.